Learn More
Biomatik Corporation Mouse Acetyl Coenzyme A Carboxylase Alpha (ACACa) ELISA Kit
Supplier:  Biomatik Corporation EKU02072
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q5SWU9. Target Synonyms: ACAC; ACC1; ACCA; ACAC; ACC2; ACCB; ACACB; ACACA; Acetyl CoA Carboxylase; Biotin carboxylase. Restriction: For Research Use Only.
Catalog No. 50-229-2051