Learn More
Biomatik Corporation Human Adenylate Cyclase 1, Brain (ADCY1) ELISA Kit
Supplier:  Biomatik Corporation EKU02136
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.31ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q08828. Target Synonyms: AC1; ATP pyrophosphate-lyase 1; Ca(2+)/calmodulin-activated adenylyl cyclase; Adenylate cyclase type I. Restriction: For Research Use Only.
Catalog No. 50-229-2061