Learn More
Biomatik Corporation Human Adenylate Cyclase 5 (ADCY5) ELISA Kit
Supplier:  Biomatik Corporation EKU02140
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O95622. Target Synonyms: Adenylate cyclase type V; ATP pyrophosphate-lyase 5. Restriction: For Research Use Only.
Catalog No. 50-229-2063