Learn More
Biomatik Corporation Human Adrenergic Receptor Alpha 1A (ADRa1A) ELISA Kit
Supplier:  Biomatik Corporation EKU02165
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P35348. Target Synonyms: ADR-A1A; ADRA1-A; ADRA1C; ADRA1L1; Alpha1AAR; Alpha 1a-Adrenoceptor. Restriction: For Research Use Only.
Catalog No. 50-229-2067