Learn More
Biomatik Corporation Mouse Adrenergic Receptor Beta 2 (ADRb2) ELISA Kit
Supplier:  Biomatik Corporation EKU02170
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P18762. Target Synonyms: B2AR; Beta-2 adrenoreceptor. Restriction: For Research Use Only.
Catalog No. 50-229-2069