Learn More
Biomatik Corporation Dog Adrenomedullin (ADM) ELISA Kit
Supplier:  Biomatik Corporation EKU02184
Encompass_Preferred
Dog Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.42pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: O77559. Gene ID: 403817. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2071