Learn More
Biomatik Corporation Mouse Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1) ELISA Kit
Supplier:  Biomatik Corporation EKU02223
Encompass_Preferred
Mouse Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1) ELISA Kit for detecting ALDH1A1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24549. Gene ID: 11668. Target Synonyms: ALDH11; ALDH1; ALDC; PUMB1; RALDH1; Retinaldehyde Dehydrogenase 1; Aldehyde dehydrogenase, cytosolic; Retinal dehydrogenase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2075