Learn More
Biomatik Corporation Human Aldolase C, Fructose Bisphosphate (ALDOC) ELISA Kit
Supplier:  Biomatik Corporation EKU02234
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P09972. Target Synonyms: ALDO-C; ALDC; ALD-C; Fructose-bisphosphate aldolase C; Brain-type aldolase. Restriction: For Research Use Only.
Catalog No. 50-229-2078