Learn More
Biomatik Corporation Bovine Alpha-Fetoprotein (AFP) ELISA Kit
Supplier:  Biomatik Corporation EKU02288
Encompass_Preferred
Bovine Alpha-Fetoprotein (AFP) ELISA Kit for detecting AFP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3SZ57. Gene ID: 506011. Target Synonyms: aFP; A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2086