Learn More
Biomatik Corporation Human Amiloride Sensitive Sodium Channel Subunit Alpha (SCNN1a) ELISA Kit
Supplier:  Biomatik Corporation EKU02304
Encompass_Preferred
Human Amiloride Sensitive Sodium Channel Subunit Alpha (SCNN1a) ELISA Kit for detecting SCNN1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P37088. Gene ID: 6337. Target Synonyms: ENaCa; ENaCalpha; SCNEA; SCNN1; Sodium Channel,Nonvoltage-Gated 1,Alpha; Epithelial Na (+) channel subunit alpha; Nonvoltage-gated sodium channel 1 subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2090