Learn More
Biomatik Corporation Human Aminoadipate Aminotransferase (AADAT) ELISA Kit
Supplier:  Biomatik Corporation EKU02312
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8N5Z0. Target Synonyms: KATII; KAT2; Kynurenine Aminotransferase II; L Kynurenine/Alpha Aminoadipate Aminotransferase; Kynurenine aminotransferase II. Restriction: For Research Use Only.
Catalog No. 50-229-2092