Learn More
Biomatik Corporation Human Amyloid Precursor Protein (APP) ELISA Kit
Supplier:  Biomatik Corporation EKU02339
Encompass_Preferred
Human Amyloid Precursor Protein (APP) ELISA Kit for detecting APP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05067. Gene ID: 351. Target Synonyms: AAA; ABeta; ABPP; AD1; APPI; CVAP; PN2; CTFgamma; Peptidase Nexin-II Alzheimer Disease; Peptidase Nexin-II; Amyloid Beta (A4) Precursor Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2096