Learn More
Biomatik Corporation Mouse Angiopoietin Like Protein 3 (ANGPTL3) ELISA Kit
Supplier:  Biomatik Corporation EKU02372
Encompass_Preferred
Mouse Angiopoietin Like Protein 3 (ANGPTL3) ELISA Kit for detecting ANGPTL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R182. Gene ID: 30924. Target Synonyms: ANGPT5; ANG-5; Angiopoietin 5; Angiopoietin-related protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2104