Learn More
Biomatik Corporation Rabbit Angiotensin I Converting Enzyme (ACE) ELISA Kit
Supplier:  Biomatik Corporation EKU02398
Encompass_Preferred
Rabbit Angiotensin I Converting Enzyme (ACE) ELISA Kit for detecting ACE in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12822. Gene ID: 100009274. Target Synonyms: CD143; ACE1; DCP1; ACEI; ACE-I; Kininase II; Angiotensin-Converting Enzyme; Peptidyl-Dipeptidase A; Dipeptidyl Carboxypeptidase 1; Angiotensin-converting enzyme, soluble form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2107