Learn More
Biomatik Corporation Rat Apolipoprotein A2 (APOA2) ELISA Kit
Supplier:  Biomatik Corporation EKU02490
Encompass_Preferred
Rat Apolipoprotein A2 (APOA2) ELISA Kit for detecting APOA2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04638. Gene ID: 25649. Target Synonyms: Apo-A2; Apolipoprotein A-II; ProapoA-II; Truncated apolipoprotein A-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2121