Learn More
Biomatik Corporation Rat Apolipoprotein A4 (APOA4) ELISA Kit
Supplier:  Biomatik Corporation EKU02494
Encompass_Preferred
Rat Apolipoprotein A4 (APOA4) ELISA Kit for detecting APOA4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 7.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02651. Gene ID: 25080. Target Synonyms: ApoA-IV; ApoAIV; Apo-A4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2123