Learn More
Biomatik Corporation Mouse Ataxia Telangiectasia Mutated (ATM) ELISA Kit
Supplier:  Biomatik Corporation EKU02602
Encompass_Preferred
Mouse Ataxia Telangiectasia Mutated (ATM) ELISA Kit for detecting ATM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62388. Gene ID: 11920. Target Synonyms: AT1; ATD; ATA; ATC; ATDC; ATE; TEL1; TELO1; Serine-protein kinase ATM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2138