Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter A3 (ABCA3) ELISA Kit
Supplier:  Biomatik Corporation EKU02607
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q99758. Target Synonyms: ABC-A3; ABC-C; ABC3; ABC-C transporter. Restriction: For Research Use Only.
Catalog No. 50-229-2140