Learn More
Biomatik Corporation Human Autophagy Related Protein 16 Like Protein 1 (ATG16L1) ELISA Kit
Supplier:  Biomatik Corporation EKU02651
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q676U5. Target Synonyms: WDR30. Restriction: For Research Use Only.
Catalog No. 50-229-2143