Learn More
Biomatik Corporation Rat AXL Receptor Tyrosine Kinase (AXL) ELISA Kit
Supplier:  Biomatik Corporation EKU02655
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 2.4-150ng/mL. Sensitivity: 0.8ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8VIA0. Target Synonyms: UFO; JTK11; Tyrosine-protein kinase receptor UFO. Restriction: For Research Use Only.
Catalog No. 50-229-2145