Learn More
Biomatik Corporation Human Beta-1,3-Glucuronyltransferase 1 (b3GAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU02689
Encompass_Preferred
Human Beta-1,3-Glucuronyltransferase 1 (b3GAT1) ELISA Kit for detecting b3GAT1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P2W7. Gene ID: 27087. Target Synonyms: CD57; B3-GAT1; GLCATP; GlcAT-P; GlcUAT-P; HNK-1; LEU7; NK-1; Glucuronosyltransferase P; Galactosylgalactosylxylosylprotein 3-Beta-Glucuronosyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2151