Learn More
Biomatik Corporation Human Beta-Lipotropic Hormone (bLPH) ELISA Kit
Supplier:  Biomatik Corporation EKU02708
Encompass_Preferred
Human Beta-Lipotropic Hormone (bLPH) ELISA Kit for detecting bLPH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.48pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01189. Gene ID: 5443. Target Synonyms: β-Lipotropin; B-LPH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2153