Learn More
Biomatik Corporation Mouse Chemokine (C-X-C motif) ligand 7 (CXCL7) ELISA Kit
Supplier:  Biomatik Corporation EKU02713
Encompass_Preferred
Mouse Chemokine (C-X-C motif) ligand 7 (CXCL7) ELISA Kit for detecting C-X-C motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.65pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQI5. Gene ID: 57349. Target Synonyms: CXCL7; PPBP; PBP; B-TG1; CTAP-III; CTAP3; CTAPIII; LA-PF4; LDGF; MDGF; NAP2; SCYB7; TC1; TC2; TGB1; THBGB1; Pro-Platelet Basic Protein; Beta-Thromboglobulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2156