Learn More
Biomatik Corporation Rat Biliverdin Reductase A (BLVRA) ELISA Kit
Supplier:  Biomatik Corporation EKU02724
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.625-40ng/mL. Sensitivity: 0.261ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P46844. Target Synonyms: BLVR; BVR; Biliverdin-IX alpha-reductase. Restriction: For Research Use Only.
Catalog No. 50-229-2161