Learn More
Biomatik Corporation Human Mitochondrial Pyruvate Carrier Protein 1 (MPC1) ELISA Kit
Supplier:  Biomatik Corporation EKU02797
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.073ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9Y5U8. Target Synonyms: CGI-129; BRP44L; Brain Protein 44 Like Protein. Restriction: For Research Use Only.
Catalog No. 50-229-2164