Learn More
Biomatik Corporation Human Breast Cancer Anti-Estrogen Resistance 3 (BCAR3) ELISA Kit
Supplier:  Biomatik Corporation EKU02806
Encompass_Preferred
Human Breast Cancer Anti-Estrogen Resistance 3 (BCAR3) ELISA Kit for detecting BCAR3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75815. Gene ID: 8412. Target Synonyms: NSP2; SH2D3B; Novel SH2-containing protein 2; SH2 domain-containing protein 3B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2166