Learn More
Biomatik Corporation Human Bromodomain Containing Protein 1 (BRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU02813
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.063ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O95696. Target Synonyms: BRL; BRPF2; BR140-like protein; Bromodomain and PHD finger-containing protein 2. Restriction: For Research Use Only.
Catalog No. 50-229-2168