Learn More
Biomatik Corporation Rat Calcineurin (CaN) ELISA Kit
Supplier:  Biomatik Corporation EKU02851
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PPP3CA; PPP3-CA; CCN1; CNA1; CALN; CALNA1; PP2B; PP2-B; Protein Phosphatase 3(formerly 2B),Catalytic Subunit,Alpha Isoformprotein Phosphatase 2B; Calcineurin A Alpha. Restriction: For Research Use Only.
Catalog No. 50-229-2172