Learn More
Biomatik Corporation Human Calcium Channel, Voltage Dependent, N-Type, Alpha 1B Subunit (CACNa1B) ELISA Kit
Supplier:  Biomatik Corporation EKU02864
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.129ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q00975. Target Synonyms: CACNN; CACNL1A5; BIII; Brain calcium channel III; Calcium channel, L type, alpha-1 polypeptide isoform 5; Voltage-gated calcium channel subunit alpha Cav2.2. Restriction: For Research Use Only.
Catalog No. 50-229-2174