Learn More
Biomatik Corporation Mouse Carboxypeptidase N1 (CPN1) ELISA Kit
Supplier:  Biomatik Corporation EKU02941
Encompass_Preferred
Mouse Carboxypeptidase N1 (CPN1) ELISA Kit for detecting CPN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JJN5. Gene ID: 93721. Target Synonyms: CPN; SCPN; ACBP; Anaphylatoxin inactivator; Arginine carboxypeptidase; Kininase-1; Lysine carboxypeptidase; Plasma carboxypeptidase B; Serum carboxypeptidase N. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2185