Learn More
Biomatik Corporation Bovine Caspase 3 (CASP3) ELISA Kit
Supplier:  Biomatik Corporation EKU02988
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Bovine. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q08DY9. Target Synonyms: CPP32; CPP32B; SCA1; Apoptain; Yama; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 3; SREBP cleavage activity 1. Restriction: For Research Use Only.
Catalog No. 50-229-2189