Learn More
Biomatik Corporation Human Caspase 5 (CASP5) ELISA Kit
Supplier:  Biomatik Corporation EKU02993
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P51878. Target Synonyms: ICE(rel)III; Cysteinyl Aspartate Specific Proteinases 5; Apoptosis-Related Cysteine Peptidase; Protease ICH-3; Protease TY. Restriction: For Research Use Only.
Catalog No. 50-229-2191