Learn More
Biomatik Corporation Mouse CCAAT/Enhancer Binding Protein Beta (CEBPb) ELISA Kit
Supplier:  Biomatik Corporation EKU03056
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 78-5,000pg/mL. Sensitivity: 28pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P28033. Target Synonyms: CRP2; IL6DBP; LAP; NF-IL6; TCF5; C/EBP-Beta; Liver-Enriched Transcriptional Activator Protein; Nuclear Factor Of Interleukin 6; Interleukin 6-Dependent DNA-Binding Protein. Restriction: For Research Use Only.
Catalog No. 50-229-2205