Learn More
Biomatik Corporation Rat Ceruloplasmin (CP) ELISA Kit
Supplier:  Biomatik Corporation EKU03083
Encompass_Preferred
Rat Ceruloplasmin (CP) ELISA Kit for detecting CP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.2ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13635. Target Synonyms: CP2; CER; Caeruloplasmin; Ferroxidase; Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2208