Learn More
Biomatik Corporation Rat Ciliary Neurotrophic Factor (CNTF) ELISA Kit
Supplier:  Biomatik Corporation EKU03194
Encompass_Preferred
Rat Ciliary Neurotrophic Factor (CNTF) ELISA Kit for detecting CNTF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 39-2,500pg/ml. Sensitivity: 17pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20294. Gene ID: 25707. Target Synonyms: HCNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2223