Learn More
Biomatik Corporation Mouse Citrate Synthase (CS) ELISA Kit
Supplier:  Biomatik Corporation EKU03197
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 1.56-100ng/mL. Sensitivity: 0.63ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9CZU6. Target Synonyms: Citrate (Si)-synthase, mitochondrial. Restriction: For Research Use Only.
Catalog No. 50-229-2225