Learn More
Biomatik Corporation Rat Cluster Of Differentiation (CD163) ELISA Kit
Supplier:  Biomatik Corporation EKU03221
Encompass_Preferred
Rat Cluster Of Differentiation (CD163) ELISA Kit for detecting CD163 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3Z9U2. Target Synonyms: M130; MM130; Scavenger receptor cysteine-rich type 1 protein M130; Hemoglobin scavenger receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2228