Learn More
Biomatik Corporation Rat Coagulation Factor IX (F9) ELISA Kit
Supplier:  Biomatik Corporation EKU03261
Encompass_Preferred
Rat Coagulation Factor IX (F9) ELISA Kit for detecting F9 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.67ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16296. Gene ID: 24946. Target Synonyms: HEMB; FIX; GLA Domain; PTC; Anti Hemophilic Factor B; Christmas Factor; Plasma Thromboplastic Component; Christmas Disease; Hemophilia B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2235