Learn More
Biomatik Corporation Human Coagulation Factor XII (F12) ELISA Kit
Supplier:  Biomatik Corporation EKU03277
Encompass_Preferred
Human Coagulation Factor XII (F12) ELISA Kit for detecting F12 in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00748. Gene ID: 2161. Target Synonyms: FXII; HAF; Contact Factor; Hageman Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2239