Learn More
Biomatik Corporation Rat Collagen Type VI Alpha 1 (COL6a1) ELISA Kit
Supplier:  Biomatik Corporation EKU03336
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.058ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: COL6-A1; OPLL; Collagen Alpha-1(VI)chain. Restriction: For Research Use Only.
Catalog No. 50-229-2247