Learn More
Biomatik Corporation Human Corticotropin Like Intermediate Lobe Peptide (CLIP) ELISA Kit
Supplier:  Biomatik Corporation EKU03466
Encompass_Preferred
Human Corticotropin Like Intermediate Lobe Peptide (CLIP) ELISA Kit for detecting CLIP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.89pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01189. Gene ID: 5443. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2260