Learn More
Biomatik Corporation Rabbit Creatine Kinase, Muscle (CKM) ELISA Kit
Supplier:  Biomatik Corporation EKU03495
Encompass_Preferred
Rabbit Creatine Kinase, Muscle (CKM) ELISA Kit for detecting CKM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.63ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00563. Gene ID: 100009056. Target Synonyms: CK-MM; CKMM; CK3; CK-M; MM-CK; M-CK; Creatine kinase M-type, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2262