Learn More
Biomatik Corporation Human Cyclin D2 (CCND2) ELISA Kit
Supplier:  Biomatik Corporation EKU03533
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1,000pg/mL. Sensitivity: 6.0pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P30279. Target Synonyms: CCN-D2; G1/S-Specific Cyclin-D2. Restriction: For Research Use Only.
Catalog No. 50-229-2270