Learn More
Biomatik Corporation Mouse Cyclin Dependent Kinase Inhibitor 2A (CDKN2A) ELISA Kit
Supplier:  Biomatik Corporation EKU03546
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.124ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P51480. Target Synonyms: P16; ARF; CDK4I; CMM2; INK4; INK4a; MLM; MTS1; TP16; P14; P14ARF; P16INK4a; P19; Melanoma P16; Cyclin-dependent kinase 4 inhibitor A; Multiple tumor suppressor 1. Restriction: For Research Use Only.
Catalog No. 50-229-2274