Learn More
Biomatik Corporation Human Cylindromatosis (CYLD) ELISA Kit
Supplier:  Biomatik Corporation EKU03555
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.118ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9NQC7. Target Synonyms: CDMT; CYLD1; CYLDI; EAC; USPL2; Turban Tumor Syndrome; Ubiquitin Carboxyl-Terminal Hydrolase CYLD; Ubiquitin Specific Peptidase Like 2; Deubiquitinating enzyme CYLD. Restriction: For Research Use Only.
Catalog No. 50-229-2276