Learn More
Biomatik Corporation Pig Cystatin C (Cys-C) ELISA Kit
Supplier:  Biomatik Corporation EKU03563
Encompass_Preferred
Pig Cystatin C (Cys-C) ELISA Kit for detecting Cys-C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 125-8,000ng/ml. Sensitivity: 55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q0Z8R0. Gene ID: 733672. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2278