Learn More
Biomatik Corporation Human Cysteine Rich Secretory Protein 3 (CRISP3) ELISA Kit
Supplier:  Biomatik Corporation EKU03576
Encompass_Preferred
Human Cysteine Rich Secretory Protein 3 (CRISP3) ELISA Kit for detecting CRISP3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.94-60ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P54108. Gene ID: 10321. Target Synonyms: CRS3; Aeg2; CRISP-3; SGP28; Specific granule protein of 28 kDa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2281