Learn More
Biomatik Corporation Human Cytochrome P450 1B1 (CYP1B1) ELISA Kit
Supplier:  Biomatik Corporation EKU03595
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.54ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q16678. Target Synonyms: CP1B; GLC3A; Cytochrome P450,Family 1,Subfamily B,Polypeptide 1; Glaucoma 3,Primary Infantile. Restriction: For Research Use Only.
Catalog No. 50-229-2284