Learn More
Biomatik Corporation Mouse Cytochrome P450 27B1 (CYP27B1) ELISA Kit
Supplier:  Biomatik Corporation EKU03599
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.625-40ng/mL. Sensitivity: 0.242ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O35084. Target Synonyms: VDD1, PDDR; CYP1, P450c1; 25-Hydroxyvitamin D3 1-Alpha-Hydroxylase; Cytochrome P450 Family 27 Subfamily B Polypeptide 1; Calcidiol 1-monooxygenase; VD3 1A hydroxylase. Restriction: For Research Use Only.
Catalog No. 50-229-2287