Learn More
Biomatik Corporation Human Defensin Beta 124 (DEFb124) ELISA Kit
Supplier:  Biomatik Corporation EKU03664
Encompass_Preferred
Human Defensin Beta 124 (DEFb124) ELISA Kit for detecting DEFb124 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NES8. Gene ID: 245937. Target Synonyms: DEFB-24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2295